Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Az084 | 929300-19-6 | 99.7% | 10 MG
SDP

Catalog No. 5000234331
Encompass_Preferred

AZ084 is a potent, selective, orally active CCR8 allosteric antagonist provided for preclinical research. The compound is characterized with manufacturer-reported purity, molecular data, solubility protocols for in vitro and in vivo use, and recommended storage conditions to support pharmacology and immunology studies.

  • Potent CCR8 allosteric antagonist with reported Ki ≈ 0.9 nM.
  • High purity reported at 99.65%.
  • Molecular weight 434.57 g/mol; formula C26H34N4O2.
  • Soluble in DMSO (250 mg/mL) and formulatable for in vivo dosing protocols.
  • Appearance: light yellow to yellow solid.
  • Storage recommendations: powder at -20°C (long term) and in solvent at -80°C for best stability.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-34331 Supplier Medchemexpress LLC Supplier No. HY11921710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZ084 is a potent, selective, orally active CCR8 allosteric antagonist provided for preclinical research. The compound is characterized with manufacturer-reported purity, molecular data, solubility protocols for in vitro and in vivo use, and recommended storage conditions to support pharmacology and immunology studies.

  • Potent CCR8 allosteric antagonist with reported Ki ≈ 0.9 nM.
  • High purity reported at 99.65%.
  • Molecular weight 434.57 g/mol; formula C26H34N4O2.
  • Soluble in DMSO (250 mg/mL) and formulatable for in vivo dosing protocols.
  • Appearance: light yellow to yellow solid.
  • Storage recommendations: powder at -20°C (long term) and in solvent at -80°C for best stability.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.