Learn More
Medchemexpress LLC Az084 | 929300-19-6 | 99.7% | 5 MG
SDP
Catalog No. 5000233596
Encompass_Preferred
AZ084 is a selective, orally active CCR8 allosteric antagonist for research use. It demonstrates sub-nanomolar potency and is supplied as a high-purity solid suitable for in vitro and in vivo pharmacology studies. Solubility and storage guidance are provided to support experimental formulation and handling.
  • Selective CCR8 allosteric antagonist with Ki ≈ 0.9 nM.
  • High purity suitable for cellular and pharmacological assays.
  • Supplied as a milligram-scale solid for in vitro and in vivo use.
  • Highly soluble in DMSO; formulation examples provided for dosing.
  • Recommended storage conditions for powder and solutions to preserve stability.
  • Intended as a research tool for immunology and inflammation studies.
Catalog No. 50-002-33596 Supplier Medchemexpress LLC Supplier No. HY1192175MG
AZ084 is a selective, orally active CCR8 allosteric antagonist for research use. It demonstrates sub-nanomolar potency and is supplied as a high-purity solid suitable for in vitro and in vivo pharmacology studies. Solubility and storage guidance are provided to support experimental formulation and handling.
  • Selective CCR8 allosteric antagonist with Ki ≈ 0.9 nM.
  • High purity suitable for cellular and pharmacological assays.
  • Supplied as a milligram-scale solid for in vitro and in vivo use.
  • Highly soluble in DMSO; formulation examples provided for dosing.
  • Recommended storage conditions for powder and solutions to preserve stability.
  • Intended as a research tool for immunology and inflammation studies.