Learn More
Selleck Chemical LLC AZ10606120 dihydrochloride S3520-5mg
SDP
Catalog No. 503109301
Encompass_Preferred
AZ10606120 dihydrochloride (2HCl) is a potent and selective antagonist for P2X7 receptor (P2X7R) with IC50 of 10 nM AZ10606120 dihydrochloride exhibits anti-depressant effects and reduces tumour growth This product is not soluble in PBS solution Please do not dissolve it in PBS for administration
Catalog No. 50-310-9301 Supplier Selleck Chemical LLC Supplier No. S35205MG
AZ10606120 dihydrochloride (2HCl) is a potent and selective antagonist for P2X7 receptor (P2X7R) with IC50 of 10 nM AZ10606120 dihydrochloride exhibits anti-depressant effects and reduces tumour growth This product is not soluble in PBS solution Please do not dissolve it in PBS for administration