Learn More
Medchemexpress LLC AZ12601011 | 2748337-86-0 | 99.8% | 100 MG
SDP
Catalog No. 5000231495
Encompass_Preferred
AZ12601011 is an orally active, selective TGFBR1 (ALK5) kinase inhibitor intended for laboratory research. It inhibits SMAD2 phosphorylation (IC50 = 18 nM; Kd = 2.9 nM), has demonstrated antitumor activity in preclinical models, and is provided with high analytical purity for biochemical assays.
  • Selective TGFBR1 inhibition with low-nanomolar potency.
  • Inhibits SMAD2 phosphorylation in signaling assays.
  • High analytical purity suitable for biochemical studies.
  • Supplied as a 100 mg solid for convenient dosing.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-31495 Supplier Medchemexpress LLC Supplier No. HY122856100MG
AZ12601011 is an orally active, selective TGFBR1 (ALK5) kinase inhibitor intended for laboratory research. It inhibits SMAD2 phosphorylation (IC50 = 18 nM; Kd = 2.9 nM), has demonstrated antitumor activity in preclinical models, and is provided with high analytical purity for biochemical assays.
  • Selective TGFBR1 inhibition with low-nanomolar potency.
  • Inhibits SMAD2 phosphorylation in signaling assays.
  • High analytical purity suitable for biochemical studies.
  • Supplied as a 100 mg solid for convenient dosing.
  • For research use only; not for human or clinical use.