Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Az12601011 | 2748337-86-0 | 99.8% | 50 MG
SDP

Catalog No. 5000230495
Encompass_Preferred

AZ12601011 is an orally active, selective TGFBR1 kinase inhibitor (also inhibits ALK4 and ALK7) used as a research reagent to study TGF-β signaling and tumor models. It inhibits SMAD2 phosphorylation with reported potency (IC50 18 nM; Kd 2.9 nM) and has shown antitumor activity in vivo. High-purity powder and DMSO stock solutions are available.

  • Selective TGFBR1, ALK4, and ALK7 inhibition.
  • Biochemical potency: IC50 18 nM; Kd 2.9 nM.
  • High purity (99.78%).
  • Available as powder and DMSO solution for in vitro studies.
  • Recommended storage: powder -20°C for long term, and solutions -80°C.

Catalog No. 50-002-30495 Supplier Medchemexpress LLC Supplier No. HY12285650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZ12601011 is an orally active, selective TGFBR1 kinase inhibitor (also inhibits ALK4 and ALK7) used as a research reagent to study TGF-β signaling and tumor models. It inhibits SMAD2 phosphorylation with reported potency (IC50 18 nM; Kd 2.9 nM) and has shown antitumor activity in vivo. High-purity powder and DMSO stock solutions are available.

  • Selective TGFBR1, ALK4, and ALK7 inhibition.
  • Biochemical potency: IC50 18 nM; Kd 2.9 nM.
  • High purity (99.78%).
  • Available as powder and DMSO solution for in vitro studies.
  • Recommended storage: powder -20°C for long term, and solutions -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.