Learn More
Medchemexpress LLC Az13705339 hemihydrate | 00-00-0 | 99.8% | 638.77 g/mol | C33H36FN7O3S·1/2H2O | 5 MG
SDP
Catalog No. 5000264511
Encompass_Preferred
AZ13705339 hemihydrate is a potent, selective PAK1 inhibitor supplied for research use. It demonstrates sub-nanomolar binding and low-nanomolar cellular activity, and is provided as a high-purity solid hemihydrate or as a 10 mM solution in DMSO with recommended storage conditions for long-term stability.
  • High purity of 99.8%.
  • Potent PAK1 inhibition with IC50 ≈ 0.33 nM.
  • Sub-nanomolar binding affinity to PAK1 and strong affinity to PAK2.
  • Available as solid hemihydrate and as a 10 mM DMSO solution.
  • DMSO solubility up to 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-64511 Supplier Medchemexpress LLC Supplier No. HY120940A5MG
AZ13705339 hemihydrate is a potent, selective PAK1 inhibitor supplied for research use. It demonstrates sub-nanomolar binding and low-nanomolar cellular activity, and is provided as a high-purity solid hemihydrate or as a 10 mM solution in DMSO with recommended storage conditions for long-term stability.
  • High purity of 99.8%.
  • Potent PAK1 inhibition with IC50 ≈ 0.33 nM.
  • Sub-nanomolar binding affinity to PAK1 and strong affinity to PAK2.
  • Available as solid hemihydrate and as a 10 mM DMSO solution.
  • DMSO solubility up to 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).