Learn More
Medchemexpress LLC AZ13705339 hemihydrate | >99.77% | 638.77 | 10 MG
SDP
Catalog No. 5000436200
Encompass_Preferred
AZ13705339 hemihydrate is a highly potent and selective PAK1 inhibitor. It demonstrates strong inhibitory activity against PAK1 and PPAK1, and high binding affinities to PAK1 and PAK2. This compound is utilized in various research applications, particularly within the field of cancer studies.
  • Highly potent and selective PAK1 inhibitor
  • Exhibits IC50s of 0.33 nM for PAK1 and 59 nM for PPAK1
  • Shows binding affinities to PAK1 and PAK2, with Kds of 0.28 nM and 0.32 nM, respectively
  • Can be utilized in cancer research
  • Inhibits α4β1-controlled adhesion
  • Prevents Siglec-8 engagement-induced eosinophil death
Catalog No. 50-004-36200 Supplier Medchemexpress LLC Supplier No. HY120940A10MG
AZ13705339 hemihydrate is a highly potent and selective PAK1 inhibitor. It demonstrates strong inhibitory activity against PAK1 and PPAK1, and high binding affinities to PAK1 and PAK2. This compound is utilized in various research applications, particularly within the field of cancer studies.
  • Highly potent and selective PAK1 inhibitor
  • Exhibits IC50s of 0.33 nM for PAK1 and 59 nM for PPAK1
  • Shows binding affinities to PAK1 and PAK2, with Kds of 0.28 nM and 0.32 nM, respectively
  • Can be utilized in cancer research
  • Inhibits α4β1-controlled adhesion
  • Prevents Siglec-8 engagement-induced eosinophil death