Learn More
AdipoGen AZ191 (25 mg)
SDP
Supplier:  AdipoGen AGCR13657M025
Encompass_Preferred
Chemical. CAS: 1594092-37-1. Formula: C24H27N7O. MW: 429.5. Synthetic. Cell permeable azaindole that inhibits the serine/threonine kinase activity of DYRK1B (IC50=17nM) with 5-fold and 110-fold selectivity against the related family members DYRK1A and DYRK2, respectively. Used as a probe to elucidate the mechanism of DYRK1B regulation of cell cycle progression. Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B (DYRK1B) belongs to a family of nuclear-localized protein kinases and participates in the regulation of the cell cycle with key roles in proliferation and differentiation. It is upregulated in solid tumors and gain-of-function mutations in the gene encoding this kinase have been linked to metabolic syndrome.
Catalog No. 50-205-4036