Learn More
AdipoGen AZ191
SDP
Catalog No. 502562249
Encompass_Preferred
Chemical. CAS 1594092-37-1. Formula C24H27N7O. MW 429.5. Synthetic. Cell permeable azaindole that inhibits the serine/threonine kinase activity of DYRK1B IC50=17nM with 5-fold and 110-fold selectivity against the related family members DYRK1A and DYRK2, respectively. Used as a probe to elucidate the mechanism of DYRK1B regulation of cell cycle progression. Dual-specificity tyrosine-Y-phosphorylation regulated kinase 1B DYRK1B belongs to a family of nuclear-localized protein kinases and participates in the regulation of the cell cycle with key roles in proliferation and differentiation. It is upregulated in solid tumors and gain-of-function mutations in the gene encoding this kinase have been linked to metabolic syndrome.
Catalog No. 50-256-2249 Supplier AdipoGen Supplier No. AGCR13657M005
Chemical. CAS 1594092-37-1. Formula C24H27N7O. MW 429.5. Synthetic. Cell permeable azaindole that inhibits the serine/threonine kinase activity of DYRK1B IC50=17nM with 5-fold and 110-fold selectivity against the related family members DYRK1A and DYRK2, respectively. Used as a probe to elucidate the mechanism of DYRK1B regulation of cell cycle progression. Dual-specificity tyrosine-Y-phosphorylation regulated kinase 1B DYRK1B belongs to a family of nuclear-localized protein kinases and participates in the regulation of the cell cycle with key roles in proliferation and differentiation. It is upregulated in solid tumors and gain-of-function mutations in the gene encoding this kinase have been linked to metabolic syndrome.