Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Az194 | 2241651-99-8 | 99.8% | 567.63 | C34H31F2N3O3 | 5 MG
SDP

Catalog No. 5000221640
Encompass_Preferred

AZ194 is a research small-molecule inhibitor of the CRMP2-Ubc9 interaction and the NaV1.7 sodium channel (IC50 = 1.2 μM). It blocks SUMOylation of CRMP2 and selectively reduces NaV1.7 surface expression, producing antinociceptive effects in preclinical pain models. Supplied as a white to off-white solid for laboratory research use.

  • Inhibits CRMP2-Ubc9 interaction and NaV1.7 (IC50 = 1.2 μM).
  • Molecular weight: 567.63 g/mol.
  • Chemical formula: C34H31F2N3O3.
  • High purity (>99.8%) suitable for research applications.
  • Appearance: white to off-white solid.
  • Soluble in DMSO at approximately 33.33 mg/mL; may require ultrasonic assistance.
  • Storage recommendations: powder at -20°C or 4°C; aliquot solutions and store at -80°C.

Catalog No. 50-002-21640 Supplier Medchemexpress LLC Supplier No. HY1451695MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZ194 is a research small-molecule inhibitor of the CRMP2-Ubc9 interaction and the NaV1.7 sodium channel (IC50 = 1.2 μM). It blocks SUMOylation of CRMP2 and selectively reduces NaV1.7 surface expression, producing antinociceptive effects in preclinical pain models. Supplied as a white to off-white solid for laboratory research use.

  • Inhibits CRMP2-Ubc9 interaction and NaV1.7 (IC50 = 1.2 μM).
  • Molecular weight: 567.63 g/mol.
  • Chemical formula: C34H31F2N3O3.
  • High purity (>99.8%) suitable for research applications.
  • Appearance: white to off-white solid.
  • Soluble in DMSO at approximately 33.33 mg/mL; may require ultrasonic assistance.
  • Storage recommendations: powder at -20°C or 4°C; aliquot solutions and store at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.