Learn More
Selleck Chemical LLC AZ20 5mg 1233339-22-4
SDP
Supplier:  Selleck Chemical LLC S70505MG
Encompass_Preferred
AZ20 is a novel potent and selective inhibitor of ATR kinase with IC50 of 5 nM in a cell-free assay, 8-fold selectivity over mTOR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4354