Learn More
Selleck Chemical LLC AZ304 S8755-5MG
SDP
Catalog No. 5000024060
Encompass_Preferred
AZ304 is a synthetic inhibitor designed to interact with the ATP-binding site of wild type and V600E mutant BRAF with IC50 values of 79 nM and 38 nM respectively It also inhibits CRAF p38 and CSF1R at sub 100 nM potencies
Catalog No. 50-000-24060 Supplier Selleck Chemical LLC Supplier No. S87555MG
AZ304 is a synthetic inhibitor designed to interact with the ATP-binding site of wild type and V600E mutant BRAF with IC50 values of 79 nM and 38 nM respectively It also inhibits CRAF p38 and CSF1R at sub 100 nM potencies