Learn More
Medchemexpress LLC HY-14710 10mg Medchemexpress, AZ3146 CAS:1124329-14-1 Purity:>98%
SDP
Catalog No. 502081630
Encompass_Preferred
Medchemexpress, HY-14710 10mg AZ3146 CAS:1124329-14-1 AZ3146 is a reasonably potent and selective Mps1 inhibitor with IC50 of 35 nM for Mps1Cat. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1630 Supplier Medchemexpress LLC Supplier No. HY1471010MG
Medchemexpress, HY-14710 10mg AZ3146 CAS:1124329-14-1 AZ3146 is a reasonably potent and selective Mps1 inhibitor with IC50 of 35 nM for Mps1Cat. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.