Learn More
Medchemexpress LLC HY-112305 5mg Medchemexpress, AZ32 CAS: Purity:>98%
SDP
Catalog No. 501969078
Encompass_Preferred
Medchemexpress, HY-112305 5mg AZ32 CAS: AZ32 is an orally bioavailable and blood-brain barrier-penetrating ATM inhibitor with an IC50 of <6.2 nM for ATM enzyme, and an IC50 of 0.31 μM for ATM in cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9078 Supplier Medchemexpress LLC Supplier No. HY1123055MG
Medchemexpress, HY-112305 5mg AZ32 CAS: AZ32 is an orally bioavailable and blood-brain barrier-penetrating ATM inhibitor with an IC50 of <6.2 nM for ATM enzyme, and an IC50 of 0.31 μM for ATM in cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.