Learn More
Selleck Chemical LLC AZ32 5mg
SDP
Supplier:  Selleck Chemical LLC S87295MG
Encompass_Preferred
AZ32 is a specific inhibitor of the ATM kinase that possesses good BBB penetration in mouse with an IC50 value of <0.0062 ?M for ATM enzyme. It shows adequate selectivity over ATR and also has high cell permeability. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8239