Learn More
Selleck Chemical LLC AZ3451-5mg
SDP
Catalog No. 502389946
Encompass_Preferred
AZ3451 is a potent antagonist of protease-activated receptor 2 (PAR2) that binds to a remote allosteric site outside the helical bundle with IC50 of 23 nM.
Catalog No. 50-238-9946 Supplier Selleck Chemical LLC Supplier No. S97445MG
AZ3451 is a potent antagonist of protease-activated receptor 2 (PAR2) that binds to a remote allosteric site outside the helical bundle with IC50 of 23 nM.