Learn More
TARGETMOL CHEMICALS INC AZ3976 5MG
SDP
Catalog No. 502580847
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ3976 is an inhibitor of PAI-1 with an IC50 of 16 uM in a plasma clot lysis assay. AZ3976 displays profibrinolytic activities. purity: 98%
Catalog No. 50-258-0847 Supplier TARGETMOL CHEMICALS INC Supplier No. T237645MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ3976 is an inhibitor of PAI-1 with an IC50 of 16 uM in a plasma clot lysis assay. AZ3976 displays profibrinolytic activities. purity: 98%