Learn More
Apexbio Technology LLC AZ505 ditrifluoroacetate,2mg CAS# 1035227-44-1
SDP
Catalog No. 502240220
Encompass_Preferred
AZ505 is a potent SET and MYND domain-containing 2 protein (SMYD2) inhibitor that selectively inhibits the enzymatic activity of SMYD2 (IC50: 0.12 μM) over a variety of other protein lysine methyltransferases (IC50 > 83.3 μΜ), including SMYD3, DOT1L, EZH2, GLP, G9A and SET7/9 [1]. AZ505 contains three distinct moieties: the benzooxazinone group which interacts with the lysine-binding channel of SMYD2; the cyclohexyl group which interacts with the core SET and I-SET domains of SMYD2; and the dichlorophenethyl group which extends across the peptide binding groove of SMYD2 [1]. The X-ray crystallographic analysis has revealed that AZ505 binds into the peptide binding groove of SMYD2 competing with the peptide substrates [1].Reference Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0220 Supplier Apexbio Technology LLC Supplier No. B12562
AZ505 is a potent SET and MYND domain-containing 2 protein (SMYD2) inhibitor that selectively inhibits the enzymatic activity of SMYD2 (IC50: 0.12 μM) over a variety of other protein lysine methyltransferases (IC50 > 83.3 μΜ), including SMYD3, DOT1L, EZH2, GLP, G9A and SET7/9 [1]. AZ505 contains three distinct moieties: the benzooxazinone group which interacts with the lysine-binding channel of SMYD2; the cyclohexyl group which interacts with the core SET and I-SET domains of SMYD2; and the dichlorophenethyl group which extends across the peptide binding groove of SMYD2 [1]. The X-ray crystallographic analysis has revealed that AZ505 binds into the peptide binding groove of SMYD2 competing with the peptide substrates [1].Reference Other sizes are also available. Please inqury us for quote.