Learn More
Medchemexpress LLC Az506 | 2043321-54-4 | 99.7% | C33H40N6O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13482810MG
Encompass_Preferred
AZ506 is a small-molecule inhibitor of the lysine methyltransferase SMYD2. It shows potent biochemical activity (SMYD2 IC50 = 17 nM) and reduces SMYD2-mediated p53 methylation in cells (cellular IC50 = 2.43 μM). The compound is supplied as solids in multiple sizes and as a 10 mM solution in DMSO for biochemical and cellular research applications.
  • Potent SMYD2 inhibition with low nanomolar biochemical potency.
  • Demonstrated cellular activity reducing p53 methylation.
  • High purity suitable for research applications (99.7%).
  • Available as solids in multiple sizes and as a ready-to-use DMSO solution.
  • Provided with safety and data documentation for laboratory use.
Catalog No. 50-002-59779