Learn More
Medchemexpress LLC AZ7550 Mesylate | 2319837-99-3 | 98.7% | 100 MG
SDP
Catalog No. 5000409333
Encompass_Preferred
AZ7550 Mesylate is an active metabolite of AZD9291 that inhibits IGF1R activity with an IC50 of 1.6 μM. This product is intended for research use only.
  • Inhibits IGF1R with an IC50 of 1.6 μM.
  • Shows potency and selectivity profile similar to AZD9291.
  • Inhibits DM cell line H1975, AM cell line PC9, and WT cell line LoVo.
  • Inhibits DM antiproliferative cell line H1975, AM cell line PC9, and WT antiproliferative cell line Calu3.
Catalog No. 50-004-09333 Supplier Medchemexpress LLC Supplier No. HYB0794B100MG
AZ7550 Mesylate is an active metabolite of AZD9291 that inhibits IGF1R activity with an IC50 of 1.6 μM. This product is intended for research use only.
  • Inhibits IGF1R with an IC50 of 1.6 μM.
  • Shows potency and selectivity profile similar to AZD9291.
  • Inhibits DM cell line H1975, AM cell line PC9, and WT cell line LoVo.
  • Inhibits DM antiproliferative cell line H1975, AM cell line PC9, and WT antiproliferative cell line Calu3.