Learn More
Medchemexpress LLC AZ7550 Mesylate | 2319837-99-3 | 98.7% | 5 MG
SDP
Catalog No. 501970125
Encompass_Preferred
AZ7550 Mesylate is an active metabolite of AZD9291 and acts as an inhibitor of IGF1R with an IC50 of 1.6 μM. This compound exhibits a potency and selectivity profile similar to its parent compound, AZD9291, making it suitable for various research applications.
  • Inhibits IGF1R with an IC50 of 1.6 μM
  • Active metabolite of AZD9291
  • Exhibits similar potency and selectivity profile to AZD9291
  • Inhibits double mutant cell line H1975 with IC50 of 45 nM
  • Inhibits activating mutant cell line PC9 with IC50 of 26 nM
  • Inhibits wild type cell line LoVo with IC50 of 786 nM
  • For research use only
Catalog No. 50-197-0125 Supplier Medchemexpress LLC Supplier No. HYB0794B5MG
AZ7550 Mesylate is an active metabolite of AZD9291 and acts as an inhibitor of IGF1R with an IC50 of 1.6 μM. This compound exhibits a potency and selectivity profile similar to its parent compound, AZD9291, making it suitable for various research applications.
  • Inhibits IGF1R with an IC50 of 1.6 μM
  • Active metabolite of AZD9291
  • Exhibits similar potency and selectivity profile to AZD9291
  • Inhibits double mutant cell line H1975 with IC50 of 45 nM
  • Inhibits activating mutant cell line PC9 with IC50 of 26 nM
  • Inhibits wild type cell line LoVo with IC50 of 786 nM
  • For research use only