Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AZ7550 mesylate (AZ7550 trimesylate salt) | 2319837-99-3 | 98.7% | 773.90 g·mol⁻¹ | C30H43N7O11S3 | 10 MG
SDP

Catalog No. 5000298205
Encompass_Preferred

AZ7550 mesylate is the mesylate (trimesylate) salt of AZ7550, an active metabolite of AZD9291 that inhibits IGF1R (IC50 = 1.6 μM). It is provided as an analytical standard for biochemical and cellular research and is offered in small milligram packages and solution formats for assay use.

  • Mesylate (trimesylate) salt form for improved handling and solubility.
  • Reported IGF1R inhibitory activity with IC50 = 1.6 μM.
  • High purity (~98.7%), suitable as an analytical standard.
  • Molecular weight 773.90 g·mol⁻¹ and formula C30H43N7O11S3.
  • Available in small quantities (5-100 mg) and as DMSO solutions for assay readiness.

Catalog No. 50-002-98205 Supplier Medchemexpress LLC Supplier No. HYB0794B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

AZ7550 mesylate is the mesylate (trimesylate) salt of AZ7550, an active metabolite of AZD9291 that inhibits IGF1R (IC50 = 1.6 μM). It is provided as an analytical standard for biochemical and cellular research and is offered in small milligram packages and solution formats for assay use.

  • Mesylate (trimesylate) salt form for improved handling and solubility.
  • Reported IGF1R inhibitory activity with IC50 = 1.6 μM.
  • High purity (~98.7%), suitable as an analytical standard.
  • Molecular weight 773.90 g·mol⁻¹ and formula C30H43N7O11S3.
  • Available in small quantities (5-100 mg) and as DMSO solutions for assay readiness.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.