Learn More
Selleck Chemical LLC AZ876
SDP
Catalog No. 5000002991
Encompass_Preferred
AZ876 is a novel high-affinity Liver X Receptor (LXR) agonist with Ki values of 0 007 M and 0 011 M for human LXR and LXR respectively
Catalog No. 50-000-02991 Supplier Selleck Chemical LLC Supplier No. S64272MG
AZ876 is a novel high-affinity Liver X Receptor (LXR) agonist with Ki values of 0 007 M and 0 011 M for human LXR and LXR respectively