Learn More
Medchemexpress LLC HY-10411 1mg , AZ960 CAS:905586-69-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104111MG
Encompass_Preferred
HY-10411 1mg AZ960 CAS: 905586-69-8 AZ960 is a potent and specific inhibitor of the JAK2 kinase with a Ki of 0.45 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9617