Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aza-cybz | 95.2% | 802.23 | C43H49ClIN3O2 | 10 MG
SDP

Catalog No. 5000297569
Encompass_Preferred

Aza-CyBz is a NaCl-responsive near-infrared fluorescent probe used to detect and visualize salt-induced stress in plant samples. It forms ordered J-aggregates with NaCl through electrostatic interaction, producing near-infrared fluorescence quenching with a reported detection limit of 170 μM. Supplied for research use, the compound is provided in milligram-scale quantities for laboratory studies.

  • Responds selectively to NaCl via J-aggregate formation.
  • Near-infrared fluorescence quenching enables imaging of salt stress.
  • Detection limit approximately 170 μM.
  • Molecular formula C43H49ClIN3O2; molecular weight 802.23.
  • Purity approximately 95.2% as supplied.
  • Store sealed at 4°C, protected from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-97569 Supplier Medchemexpress LLC Supplier No. HYD286410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aza-CyBz is a NaCl-responsive near-infrared fluorescent probe used to detect and visualize salt-induced stress in plant samples. It forms ordered J-aggregates with NaCl through electrostatic interaction, producing near-infrared fluorescence quenching with a reported detection limit of 170 μM. Supplied for research use, the compound is provided in milligram-scale quantities for laboratory studies.

  • Responds selectively to NaCl via J-aggregate formation.
  • Near-infrared fluorescence quenching enables imaging of salt stress.
  • Detection limit approximately 170 μM.
  • Molecular formula C43H49ClIN3O2; molecular weight 802.23.
  • Purity approximately 95.2% as supplied.
  • Store sealed at 4°C, protected from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.