Learn More
Medchemexpress LLC HY-136383 5mg Medchemexpress, AZA1 CAS:1071098-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1363835MG
Encompass_Preferred
Medchemexpress, HY-136383 5mg AZA1 CAS:1071098-42-4 AZA1 is a potent dual inhibitor of Rac1 and Cdc42. AZA1 induces prostate cancer cells apoptosis and inhibits prostate cancer cells proliferation, migration and invasion[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0432