Learn More
Selleck Chemical LLC Azacitidine (5-Azacytidine) 50mg 320-67-2 5-AzaC,Ladakamycin, AZA,5-Aza
SDP
Catalog No. NC0596135
Encompass_Preferred
Azacitidine (5-Azacytidine, 5-AzaC, Ladakamycin, AZA, 5-Aza) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation by trapping DNA methyltransferases. Azacitidine induces mitochondrial apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0596135 Supplier Selleck Chemical LLC Supplier No. S178250MG
Add to Cart
Add to Cart
Azacitidine (5-Azacytidine, 5-AzaC, Ladakamycin, AZA, 5-Aza) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation by trapping DNA methyltransferases. Azacitidine induces mitochondrial apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.