Learn More
Cayman Chemical Azacyclonol, Purity: ≥95%, CAS: 115-46-8, 25 g
SDP
Catalog No. 502939258
Encompass_Preferred
A CNS depressant; reduces transmission through the sympathetic ganglia, decreasing electrically stimulated contractile responses in feline nictitating membrane; reduces coordinated locomotor activity in mice by >50% in mice at 71, 142, and 213 mg/kg; decreases hyperactivity induced by pipradol, D-amphetamine, morphine, and cocaine; increases the duration of hexobarbital-induced sleeping time; a metabolite of terfenadine formed via CYP3A4
Catalog No. 50-293-9258 Supplier Cayman Chemical Supplier No. 2730325 G
A CNS depressant; reduces transmission through the sympathetic ganglia, decreasing electrically stimulated contractile responses in feline nictitating membrane; reduces coordinated locomotor activity in mice by >50% in mice at 71, 142, and 213 mg/kg; decreases hyperactivity induced by pipradol, D-amphetamine, morphine, and cocaine; increases the duration of hexobarbital-induced sleeping time; a metabolite of terfenadine formed via CYP3A4