Learn More
Medchemexpress LLC HY-B0530 500mg Medchemexpress, Azacyclonol CAS:115-46-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0530500MG
Encompass_Preferred
Medchemexpress, HY-B0530 500mg Azacyclonol CAS:115-46-8 Azacyclonol (γ-pipradol), a metabolite of Terfenadine, is a central depressant agent. Azacyclonol is a ganglion-blocking agent. Azacyclonol can be used to diminish psychoses-induced hallucinations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2584