Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azadibenzocyclooctyne-maleimide

Catalog No. AAJ65377MB
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65377MB 25 mg
AAJ65377MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65377MB Supplier Thermo Scientific Chemicals Supplier No. J65377MB
Add to Cart

For incorporation of ADIBO functionality into sulfhydryl containing compounds | C25H21N3O4

For incorporation of ADIBO functionality into sulfhydryl containing compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C25H21N3O4
MDL Number MFCD22380750
Synonym ADIBO-maleimide

Specifications

Color White to Gray
Quantity 25 mg
Solubility Information Solulbe in dichloromethane,DMF,DMSO,or THF
Formula Weight 427.45
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-maleimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.