Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azadibenzocyclooctyne-PEG, MW 5,000

Catalog No. AAJ64977MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64977MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64977MC Supplier Thermo Scientific Chemicals Supplier No. J64977MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

 

Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym ADIBO-PEG, MW 5,000

Specifications

Color White to Gray
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 5kDa
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-PEG
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.