Learn More
Medchemexpress LLC HY-126741 1mg , Azadirachtin CAS:11141-17-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1267411MG
Encompass_Preferred
HY-126741 1mg Azadirachtin CAS: 11141-17-6 Azadirachtin, one of the most promising botanical insecticides, is widely used for pest control. Azadirachtin induces apoptosis in insect cell lines, including Sf9, SL-1 and BTI-Tn-5B1-4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1727