Learn More
Apexbio Technology LLC Azaguanine-8 134-58-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062010
Encompass_Preferred
Azaguanine-8 (CAS 134-58-7) is a synthetic purine analog utilized in biomedical research due to its antineoplastic properties Structurally similar to guanine Azaguanine-8 exerts its biological activity by competitively interfering with guanine metabolism subsequently disrupting nucleic acid synthesis and cellular replication pathways Through this mechanism the compound demonstrates inhibitory effects on cell proliferation making it a valuable tool in investigating nucleic acid metabolism and cancer biology in preclinical studies
Catalog No. 50-000-62010 Supplier Apexbio Technology LLC Supplier No. B16675.1
Azaguanine-8 (CAS 134-58-7) is a synthetic purine analog utilized in biomedical research due to its antineoplastic properties Structurally similar to guanine Azaguanine-8 exerts its biological activity by competitively interfering with guanine metabolism subsequently disrupting nucleic acid synthesis and cellular replication pathways Through this mechanism the compound demonstrates inhibitory effects on cell proliferation making it a valuable tool in investigating nucleic acid metabolism and cancer biology in preclinical studies