Learn More
Selleck Chemical LLC Azaindole 1
SDP
Supplier:  Selleck Chemical LLC S66365MG
Encompass_Preferred
Azaindole 1 (BAY-549) is a selective Rho-associated protein kinase (ROCK) inhibitor with IC50 of 0 6 and 1 1 nM for human ROCK-1 and ROCK-2 in an ATP-competitive manner
Catalog No. 50-000-00036