Learn More
Medchemexpress LLC Azalamellarin N | 1809535-57-6 | 99.6% | 498.48 | 25 MG
SDP
Catalog No. 5000392634
Encompass_Preferred
Azalamellarin N is an inhibitor of pyroptosis and has different inhibitory effects on different pyroptosis inducers. It inhibits pyroptosis by targeting molecules that act upstream of NLRP3 inflammasome activation, rather than directly targeting components of the NLRP3 inflammasome.
  • Inhibits pyroptosis by targeting molecules upstream of NLRP3 inflammasome activation.
  • Does not directly target components of the NLRP3 inflammasome.
  • Inhibitory potency against different pyroptosis inducers: nigericin > R837.
Catalog No. 50-003-92634 Supplier Medchemexpress LLC Supplier No. 25MGHY162150
Azalamellarin N is an inhibitor of pyroptosis and has different inhibitory effects on different pyroptosis inducers. It inhibits pyroptosis by targeting molecules that act upstream of NLRP3 inflammasome activation, rather than directly targeting components of the NLRP3 inflammasome.
  • Inhibits pyroptosis by targeting molecules upstream of NLRP3 inflammasome activation.
  • Does not directly target components of the NLRP3 inflammasome.
  • Inhibitory potency against different pyroptosis inducers: nigericin > R837.