Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azaleatin | 529-51-1 | 5 MG
SDP

Catalog No. 5000419590
Encompass_Preferred

Azaleatin is an O-methylated flavonol isolated from Rhododendron species. It functions as a dipeptidyl peptidase-IV inhibitor and is utilized in research for type-2 diabetes and obesity.

  • Isolated from rhododendron species
  • Functions as a dipeptidyl peptidase-IV inhibitor
  • Useful for research on type-2 diabetes
  • Useful for research on obesity
  • Purity of 98.80%
  • Molecular weight of 316.26
  • Chemical formula C16H12O7
  • Appearance as an off-white to light yellow solid
  • Classified as flavonoids, flavonols, phenols, polyphenols
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-19590 Supplier Medchemexpress LLC Supplier No. HYN76535MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azaleatin is an O-methylated flavonol isolated from Rhododendron species. It functions as a dipeptidyl peptidase-IV inhibitor and is utilized in research for type-2 diabetes and obesity.

  • Isolated from rhododendron species
  • Functions as a dipeptidyl peptidase-IV inhibitor
  • Useful for research on type-2 diabetes
  • Useful for research on obesity
  • Purity of 98.80%
  • Molecular weight of 316.26
  • Chemical formula C16H12O7
  • Appearance as an off-white to light yellow solid
  • Classified as flavonoids, flavonols, phenols, polyphenols
  • Soluble in DMSO at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.