Learn More
Medchemexpress LLC Azamethiphos 250mg | 35575-96-3 | 324.68 g/mol | C9H10ClN2O5PS | 250 MG
SDP
Catalog No. 5000229521
Encompass_Preferred
Azamethiphos analytical standard is a reference-grade organothiophosphate insecticide (CAS 35575-96-3) used as a characterized material for analytical, calibration, and research applications, including method development and quantitative assays.
  • Analytical standard grade suitable for HPLC and GC calibration and method validation.
  • High purity (>98% by HPLC when specified) for reliable quantitation and trace analysis.
  • Appropriate for residue analysis and environmental monitoring of organophosphate compounds.
  • Available in small pack sizes, including 250 mg, to support routine lab workflows.
  • Store and handle according to the certificate of analysis and safety data sheet.
Catalog No. 50-002-29521 Supplier Medchemexpress LLC Supplier No. HY114899250MG
Azamethiphos analytical standard is a reference-grade organothiophosphate insecticide (CAS 35575-96-3) used as a characterized material for analytical, calibration, and research applications, including method development and quantitative assays.
  • Analytical standard grade suitable for HPLC and GC calibration and method validation.
  • High purity (>98% by HPLC when specified) for reliable quantitation and trace analysis.
  • Appropriate for residue analysis and environmental monitoring of organophosphate compounds.
  • Available in small pack sizes, including 250 mg, to support routine lab workflows.
  • Store and handle according to the certificate of analysis and safety data sheet.