Learn More
Apexbio Technology LLC Azaperone 1649-18-9 50mg
SDP
Catalog No. 502240783
Encompass_Preferred
Azaperone is a small-molecule antagonist targeting dopamine receptors particularly D2 receptors It is designed to antagonize dopaminergic neurotransmission thereby modulating neural signaling pathways Azaperone exerts its biological activity primarily through dopamine receptor antagonism Based on these pharmacological properties azaperone holds research potential in animal behavioral experiments facilitating sedation stress management and immobilization in veterinary clinical and research settings
This item is not returnable. View return policy
Catalog No. 50-224-0783 Supplier Apexbio Technology LLC Supplier No. B166850
Azaperone is a small-molecule antagonist targeting dopamine receptors particularly D2 receptors It is designed to antagonize dopaminergic neurotransmission thereby modulating neural signaling pathways Azaperone exerts its biological activity primarily through dopamine receptor antagonism Based on these pharmacological properties azaperone holds research potential in animal behavioral experiments facilitating sedation stress management and immobilization in veterinary clinical and research settings