Learn More
Medchemexpress LLC Azaperone-d4 | 1173021-72-1 | 1MG
SDP
Catalog No. 5000411160
Encompass_Preferred
Azaperone-d4 is a deuterium-labeled version of Azaperone, a dopamine antagonist that also exhibits antihistaminic and anticholinergic properties. This pyridinylpiperazine and butyrophenone neuroleptic agent is known for its antiemetic effects and is primarily used as a tranquilizer in veterinary medicine. Deuterium labeling incorporates stable heavy isotopes into drug molecules, serving as tracers for quantitation during drug development and potentially affecting pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Dopamine antagonist
  • Exhibits antihistaminic properties
  • Exhibits anticholinergic properties
  • Pyridinylpiperazine and butyrophenone neuroleptic agent
  • Provides antiemetic effects
  • Used as a tranquilizer in veterinary medicine
  • Useful as a tracer for drug development
Catalog No. 50-004-11160 Supplier Medchemexpress LLC Supplier No. HYB1470S1MG
Azaperone-d4 is a deuterium-labeled version of Azaperone, a dopamine antagonist that also exhibits antihistaminic and anticholinergic properties. This pyridinylpiperazine and butyrophenone neuroleptic agent is known for its antiemetic effects and is primarily used as a tranquilizer in veterinary medicine. Deuterium labeling incorporates stable heavy isotopes into drug molecules, serving as tracers for quantitation during drug development and potentially affecting pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Dopamine antagonist
  • Exhibits antihistaminic properties
  • Exhibits anticholinergic properties
  • Pyridinylpiperazine and butyrophenone neuroleptic agent
  • Provides antiemetic effects
  • Used as a tranquilizer in veterinary medicine
  • Useful as a tracer for drug development