Learn More
Medchemexpress LLC Azaphen dihydrochloride monohydrate | 63302-99-8 | 99.9% | C16H23Cl2N5O2 | 1 ML
SDP
Catalog No. 5000432621
Encompass_Preferred
Azaphen dihydrochloride monohydrate, also known as Pipofezine, is a potent inhibitor of serotonin reuptake. This tricyclic antidepressant (TCA) is approved in Russia for the treatment of depression.
  • Potent inhibitor of serotonin reuptake
  • Tricyclic antidepressant for depression treatment
  • Available as a 10 mM solution in water
  • High purity for reliable research applications
Catalog No. 50-004-32621 Supplier Medchemexpress LLC Supplier No. HYA0022A10MM/1ML
Azaphen dihydrochloride monohydrate, also known as Pipofezine, is a potent inhibitor of serotonin reuptake. This tricyclic antidepressant (TCA) is approved in Russia for the treatment of depression.
  • Potent inhibitor of serotonin reuptake
  • Tricyclic antidepressant for depression treatment
  • Available as a 10 mM solution in water
  • High purity for reliable research applications