Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azapropazone 5mg | 13539-59-8 | 300.36 g/mol | C16H20N4O2 | 5 MG
SDP

Catalog No. 5000227215
Encompass_Preferred

Azapropazone is a nonsteroidal anti-inflammatory drug (NSAID) supplied as an analytical research standard for non-clinical biochemical and pharmacological studies. It is used in research on rheumatoid arthritis and other inflammatory rheumatic conditions.

  • High purity, 99.7% (HPLC), suitable for analytical applications.
  • Molecular weight 300.36 g/mol and formula C16H20N4O2 for exact mass-based calculations.
  • Available in small milligram quantities for assay development and reference standards.
  • Soluble in common organic solvents for preparation of stock solutions.

Catalog No. 50-002-27215 Supplier Medchemexpress LLC Supplier No. HY1164425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azapropazone is a nonsteroidal anti-inflammatory drug (NSAID) supplied as an analytical research standard for non-clinical biochemical and pharmacological studies. It is used in research on rheumatoid arthritis and other inflammatory rheumatic conditions.

  • High purity, 99.7% (HPLC), suitable for analytical applications.
  • Molecular weight 300.36 g/mol and formula C16H20N4O2 for exact mass-based calculations.
  • Available in small milligram quantities for assay development and reference standards.
  • Soluble in common organic solvents for preparation of stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.