Learn More
Medchemexpress LLC Azasetron hydrochloride | 123040-16-4 | 99.8% | C17H21Cl2N3O3 | 1 ML
SDP
Catalog No. 5000433410
Encompass_Preferred
Azasetron hydrochloride (Y-25130 hydrochloride) is a benzamide derivative that functions as a potent and selective 5-HT3 receptor antagonist. It is currently under study for its application in treating chemotherapy-induced nausea and vomiting (CINV).
  • Potent and selective 5-HT3 receptor antagonist.
  • Used to study chemotherapy-induced nausea and vomiting (CINV).
  • Can effectively penetrate the skin and enter systemic circulation.
Catalog No. 50-004-33410 Supplier Medchemexpress LLC Supplier No. HYB006810MM/1ML
Azasetron hydrochloride (Y-25130 hydrochloride) is a benzamide derivative that functions as a potent and selective 5-HT3 receptor antagonist. It is currently under study for its application in treating chemotherapy-induced nausea and vomiting (CINV).
  • Potent and selective 5-HT3 receptor antagonist.
  • Used to study chemotherapy-induced nausea and vomiting (CINV).
  • Can effectively penetrate the skin and enter systemic circulation.