Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azasetron hydrochloride | 123040-16-4 | 99.8% | C17H21Cl2N3O3 | 100 MG
SDP

Catalog No. 5000432157
Encompass_Preferred

Azasetron hydrochloride is a benzamide derivative that functions as a potent and selective antagonist of the 5-HT3 receptor, demonstrating an IC50 of 0.33 nM against this target. It has been investigated for its potential in studies related to chemotherapy-induced nausea and vomiting (CINV) and has shown the ability to penetrate the skin and enter systemic circulation in in vivo models.

  • Mechanism of action: Potent and selective 5-HT3 receptor antagonist.
  • Target affinity: IC50 of 0.33 nM for 5-HT3 receptor.
  • Chemical classification: Benzamide derivative.
  • Molecular formula: C17H21Cl2N3O3.
  • Molecular weight: 386.27.
  • Purity: 99.76%.
  • Physical characteristics: White to off-white solid.
  • Solubility: Soluble in water (20 mg/mL) and DMSO (2.22 mg/mL).
  • Research application: Used in CINV studies.
  • Pharmacokinetic property: Penetrates skin and enters systemic circulation in animal models.

Catalog No. 50-004-32157 Supplier Medchemexpress LLC Supplier No. HYB0068100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azasetron hydrochloride is a benzamide derivative that functions as a potent and selective antagonist of the 5-HT3 receptor, demonstrating an IC50 of 0.33 nM against this target. It has been investigated for its potential in studies related to chemotherapy-induced nausea and vomiting (CINV) and has shown the ability to penetrate the skin and enter systemic circulation in in vivo models.

  • Mechanism of action: Potent and selective 5-HT3 receptor antagonist.
  • Target affinity: IC50 of 0.33 nM for 5-HT3 receptor.
  • Chemical classification: Benzamide derivative.
  • Molecular formula: C17H21Cl2N3O3.
  • Molecular weight: 386.27.
  • Purity: 99.76%.
  • Physical characteristics: White to off-white solid.
  • Solubility: Soluble in water (20 mg/mL) and DMSO (2.22 mg/mL).
  • Research application: Used in CINV studies.
  • Pharmacokinetic property: Penetrates skin and enters systemic circulation in animal models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.