Learn More
Medchemexpress LLC Azasetron hydrochloride | 123040-16-4 | MFCD00209913 | 99.8% | C17H21Cl2N3O3 | 5 MG
SDP
Catalog No. 5000432162
Encompass_Preferred
Azasetron hydrochloride is a benzamide derivative functioning as a potent and selective 5-HT3 receptor antagonist. It is utilized in studies concerning chemotherapy-induced nausea and vomiting (CINV).
  • Potent and selective 5-HT3 receptor antagonist
  • Used in studies for chemotherapy-induced nausea and vomiting (CINV)
  • Purity of 99.76%
  • Solid, white to off-white appearance
  • Effectively penetrates skin in in vivo studies
  • IC50 for 5-HT3 Receptor is 0.33 nM
Catalog No. 50-004-32162 Supplier Medchemexpress LLC Supplier No. HYB00685MG
Azasetron hydrochloride is a benzamide derivative functioning as a potent and selective 5-HT3 receptor antagonist. It is utilized in studies concerning chemotherapy-induced nausea and vomiting (CINV).
  • Potent and selective 5-HT3 receptor antagonist
  • Used in studies for chemotherapy-induced nausea and vomiting (CINV)
  • Purity of 99.76%
  • Solid, white to off-white appearance
  • Effectively penetrates skin in in vivo studies
  • IC50 for 5-HT3 Receptor is 0.33 nM