Learn More
Medchemexpress LLC Azasetron hydrochloride (Y-25130 hydrochloride) | 123040-16-4 | 99.8% | C17H21Cl2N3O3 | 50 MG
SDP
Catalog No. 5000432498
Encompass_Preferred
Azasetron hydrochloride is a potent and selective 5-HT3 receptor antagonist, a benzamide derivative used in the study of Chemotherapy-induced nausea and vomiting (CINV). This product is for research use only. It has an IC50 for the 5-HT3 Receptor of 0.33 nM and appears as a white to off-white solid with a molecular weight of 386.27.
  • Potent and selective 5-HT3 receptor antagonist
  • Used in studies for chemotherapy-induced nausea and vomiting (CINV)
  • Effectively penetrates skin and enters systemic circulation (demonstrated in in vivo studies)
  • Soluble in H2O (20 mg/mL) and DMSO (2.22 mg/mL)
  • Clear solution of 50 mg/mL in PBS achievable for in vivo dissolution
Catalog No. 50-004-32498 Supplier Medchemexpress LLC Supplier No. HYB006850MG
Azasetron hydrochloride is a potent and selective 5-HT3 receptor antagonist, a benzamide derivative used in the study of Chemotherapy-induced nausea and vomiting (CINV). This product is for research use only. It has an IC50 for the 5-HT3 Receptor of 0.33 nM and appears as a white to off-white solid with a molecular weight of 386.27.
  • Potent and selective 5-HT3 receptor antagonist
  • Used in studies for chemotherapy-induced nausea and vomiting (CINV)
  • Effectively penetrates skin and enters systemic circulation (demonstrated in in vivo studies)
  • Soluble in H2O (20 mg/mL) and DMSO (2.22 mg/mL)
  • Clear solution of 50 mg/mL in PBS achievable for in vivo dissolution