Learn More
Medchemexpress LLC Azatadine (dimaleate) | 3978-86-7 | 99.85% | 200 MG
SDP
Catalog No. 5000407249
Encompass_Preferred
Azatadine dimaleate is an inhibitor of histamine and cholinergic with an IC50 of 6. It also exhibits antiserotonin activity and can be used in research on perennial allergic rhinitis.
  • Inhibitor of histamine and cholinergic
  • Exhibits antiserotonin activity
  • Can be studied in research on perennial allergic rhinitis
  • Targets histamine receptor
  • Involves GPCR/G protein, immunology/inflammation, and neuronal signaling pathways
Catalog No. 50-004-07249 Supplier Medchemexpress LLC Supplier No. HYB0170A200MG
Azatadine dimaleate is an inhibitor of histamine and cholinergic with an IC50 of 6. It also exhibits antiserotonin activity and can be used in research on perennial allergic rhinitis.
  • Inhibitor of histamine and cholinergic
  • Exhibits antiserotonin activity
  • Can be studied in research on perennial allergic rhinitis
  • Targets histamine receptor
  • Involves GPCR/G protein, immunology/inflammation, and neuronal signaling pathways