Learn More
Apexbio Technology LLC Azathioprine 446-86-6 1g
SDP
Catalog No. 502240340
Encompass_Preferred
Azathioprine (CAS 446-86-6) is an immunosuppressive prodrug that is metabolized in vivo to 6-mercaptopurine It interferes with purine synthesis leading to the inhibition of DNA RNA and protein synthesis thereby suppressing the proliferation of T and B lymphocytes Azathioprine is widely utilized in biomedical research to study mechanisms of immune modulation graft rejection and the pathophysiology of autoimmune diseases Its ability to modulate adaptive immune responses makes it a valuable tool for investigating immunosuppressive protocols and immune homeostasis in transplantation and autoimmune disorder models
This item is not returnable. View return policy
Catalog No. 50-224-0340 Supplier Apexbio Technology LLC Supplier No. B18921000
Azathioprine (CAS 446-86-6) is an immunosuppressive prodrug that is metabolized in vivo to 6-mercaptopurine It interferes with purine synthesis leading to the inhibition of DNA RNA and protein synthesis thereby suppressing the proliferation of T and B lymphocytes Azathioprine is widely utilized in biomedical research to study mechanisms of immune modulation graft rejection and the pathophysiology of autoimmune diseases Its ability to modulate adaptive immune responses makes it a valuable tool for investigating immunosuppressive protocols and immune homeostasis in transplantation and autoimmune disorder models