Learn More
Selleck Chemical LLC Azathioprine-50mg
SDP
Catalog No. 502389710
Encompass_Preferred
Azathioprine(BW 57-322) is an immunosuppressive drug, inhibiting purine synthesis and GTP-binding protein Rac1 activation, used in the treatment of organ transplantation and autoimmune diseases.
Catalog No. 50-238-9710 Supplier Selleck Chemical LLC Supplier No. S172150MG
Azathioprine(BW 57-322) is an immunosuppressive drug, inhibiting purine synthesis and GTP-binding protein Rac1 activation, used in the treatment of organ transplantation and autoimmune diseases.