Learn More
Medchemexpress LLC Azathioprine (standard) | 446-86-6 | 99.8% | 250 MG
SDP
Catalog No. 5000406569
Encompass_Preferred
Azathioprine (Standard) is the analytical standard of Azathioprine, intended for research and analytical applications. It is an orally active immunosuppressive agent that can be converted in vivo to the active metabolite 6-mercaptopurine (6-MP). Azathioprine has myelosuppressive effects and induces apoptosis, and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for azathioprine
  • Orally active immunosuppressive agent
  • Converts to 6-mercaptopurine (6-MP) in vivo
  • Has myelosuppressive effects and induces apoptosis
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-06569 Supplier Medchemexpress LLC Supplier No. 250MGHYB0256R
Azathioprine (Standard) is the analytical standard of Azathioprine, intended for research and analytical applications. It is an orally active immunosuppressive agent that can be converted in vivo to the active metabolite 6-mercaptopurine (6-MP). Azathioprine has myelosuppressive effects and induces apoptosis, and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for azathioprine
  • Orally active immunosuppressive agent
  • Converts to 6-mercaptopurine (6-MP) in vivo
  • Has myelosuppressive effects and induces apoptosis
  • Used in HPLC, GC, and MS research experiments