Learn More
Medchemexpress LLC AZD-0364 is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.|50mg|HY-111483|99.75%|2097416-76-5| FOR RESEARCH USE ONLY.DO NOT SELL TO PATIENTS
SDP
Catalog No. NC1948535
Encompass_Preferred
MEDCHEMEXPRESS LLC|AZD-0364 is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.|50mg|HY-111483|99.75%|2097416-76-5| FOR RESEARCH USE ONLY.DO NOT SELL TO PATIENTS
Catalog No. NC1948535 Supplier Medchemexpress LLC Supplier No. HY11148350MG
MEDCHEMEXPRESS LLC|AZD-0364 is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.|50mg|HY-111483|99.75%|2097416-76-5| FOR RESEARCH USE ONLY.DO NOT SELL TO PATIENTS